Gene Transcripts

Gene Symbol
FNDC5
Entrez Gene ID
252995
Gene Name
fibronectin type III domain containing 5
Gene Aliases
FRCP2, irisin
Chromosome Location
Chr. 1: 32862268 - 32872492 on Build GRCh38
UniGene ID
Hs.524234
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001171940.1 NP_001165411.2 4, 5 559
NM_001171941.2 NP_001165412.1 4, 5 493
NM_153756.2 NP_715637.2 4, 5 559
GenBank mRNA AK092102.1 - 4, 5 492
BC062297.1 - 4, 5 482 6338770
BX537781.1 - 4, 5 435
DA393478.1 - 2, 3 163

View Full Product Details