Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001098519.1 | NP_001091989.1 | 7 | 1337 | |
NM_152759.4 | NP_689972.3 | 7 | 1226 | ||
XM_017019124.1 | XP_016874613.1 | 7 | 1345 | ||
GenBank mRNA | AK124107.1 | - | 9 | 1606 | |
BC029245.2 | - | 6 | 1161 | 5169080 |