Gene Transcripts

Gene Symbol
TSGA10IP
Entrez Gene ID
254187
Gene Name
testis specific 10 interacting protein
Gene Aliases
FAM161C
Chromosome Location
Chr. 11: 65945445 - 65960437 on Build GRCh38
UniGene ID
Hs.350671
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152762.2 NP_689975.2 4 1152
XM_011544887.2 XP_011543189.1 4 1351
XM_011544888.2 XP_011543190.1 4 1149
GenBank mRNA AK057442.1 - 4 1152
BC104022.1 - 4 1152 40025091

View Full Product Details