Gene Transcripts

Gene Symbol
OR52B2
Entrez Gene ID
255725
Gene Name
olfactory receptor family 52 subfamily B member 2
Gene Aliases
OR11-70
Chromosome Location
Chr. 11: 6169355 - 6170326 on Build GRCh38
UniGene ID
Hs.690457
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004052.1 NP_001004052.1 1 345
GenBank mRNA BC136732.1 - 1 427
BC136733.1 - 1 427 9020723

View Full Product Details