Gene Transcripts

Gene Symbol
OR5J2
Entrez Gene ID
282775
Gene Name
olfactory receptor family 5 subfamily J member 2
Gene Aliases
OR11-266
Chromosome Location
Chr. 11: 56176618 - 56177556 on Build GRCh38
UniGene ID
Hs.537145
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005492.1 NP_001005492.1 1 405
GenBank mRNA X64974.1 - 1 31

View Full Product Details