Gene Transcripts

Gene Symbol
KLK9
Entrez Gene ID
284366
Gene Name
kallikrein related peptidase 9
Gene Aliases
KLK-L3, KLKL3
Chromosome Location
Chr. 19: 51002513 - 51009634 on Build GRCh38
UniGene ID
Hs.448942
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012315.1 NP_036447.1 5 718
GenBank mRNA AY551001.1 - 4 561

View Full Product Details