Gene Transcripts

Gene Symbol
OR4D10
Entrez Gene ID
390197
Gene Name
olfactory receptor family 4 subfamily D member 10
Gene Aliases
OR11-251, OR4D10P, OST711
Chromosome Location
Chr. 11: 59477430 - 59478365 on Build GRCh38
UniGene ID
Hs.553756
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004705.1 NP_001004705.1 1 196
GenBank mRNA BC136893.1 - 1 253
BC136894.1 - 1 253 9020885

View Full Product Details