Gene Transcripts

Gene Symbol
OR8A1
Entrez Gene ID
390275
Gene Name
olfactory receptor family 8 subfamily A member 1
Gene Aliases
OR11-318, OST025
Chromosome Location
Chr. 11: 124570069 - 124571049 on Build GRCh38
UniGene ID
Hs.632104
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005194.1 NP_001005194.1 1 243
GenBank mRNA AY726592.1 - 1 119
BC137194.1 - 1 315 9021191
BC137197.1 - 1 315 9021194

View Full Product Details