Gene Transcripts

Gene Symbol
MYO18A
Entrez Gene ID
399687
Gene Name
myosin XVIIIA
Gene Aliases
MYSPDZ, SPR210
Chromosome Location
Chr. 17: 29073510 - 29180389 on Build GRCh38
UniGene ID
Hs.462590
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_078471.3 NP_510880.2 26 4187
NM_203318.1 NP_976063.1 26 4187
GenBank mRNA AB177858.1 - 25 4006
AB177860.1 - 26 3139
AK302505.1 - 25 2940
AK302725.1 - 10 901
AK303522.1 - 25 3077
AY703984.1 - 26 3139
BC039612.1 - 26 4291 6045482
CR933614.1 - 26 4162
D86970.2 - 26 4210

View Full Product Details