Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000631.4 | NP_000622.2 | 6 | 679 | |
NM_013416.3 | NP_038202.2 | 6 | 679 | ||
XM_017028808.1 | XP_016884297.1 | 6 | 1043 | ||
GenBank mRNA | AB025219.1 | - | 6 | 531 | |
AB025220.1 | - | 6 | 531 | ||
AK223324.1 | - | 6 | 650 | ||
AK290924.1 | - | 6 | 601 | ||
BC002798.1 | - | 6 | 679 | 3634177 | |
BT007346.1 | - | 6 | 495 | ||
BX355636.2 | - | 6 | 655 | ||
CR456528.1 | - | 6 | 576 | ||
CR542078.1 | - | 6 | 495 | ||
X77094.1 | - | 6 | 625 |