Gene Transcripts

Gene Symbol
PCP4
Entrez Gene ID
5121
Gene Name
Purkinje cell protein 4
Gene Aliases
PEP-19
Chromosome Location
Chr. 21: 39867421 - 39929397 on Build GRCh38
UniGene ID
Hs.80296
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006198.2 NP_006189.2 3 155
GenBank mRNA AF416716.1 - 3 103
AK289964.1 - 3 138
AM183336.1 - 3 93
BC013791.1 - 3 131 4345740
BQ287895.1 - 1 408
CR450319.1 - 2 74
U52969.1 - 3 155
X93349.1 - 3 132

View Full Product Details