Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001001329.2 | NP_001001329.1 | 4 | 742 | |
NM_001289102.1 | NP_001276031.1 | 3 | 593 | ||
NM_001289103.1 | NP_001276032.1 | 4 | 746 | ||
NM_001289104.1 | NP_001276033.1 | 4 | 742 | ||
NM_002743.3 | NP_002734.2 | 4 | 354 | ||
XM_011528130.1 | XP_011526432.1 | 5 | 508 | ||
XM_011528131.1 | XP_011526433.1 | 5 | 508 | ||
XM_011528132.1 | XP_011526434.1 | 5 | 508 | ||
XM_017026977.1 | XP_016882466.1 | 4 | 358 | ||
GenBank mRNA | AF144075.1 | - | 3 | 215 | |
AK130663.1 | - | 4 | 370 | ||
AK225279.1 | - | 4 | 368 | ||
AK225390.1 | - | 4 | 350 | ||
AK290433.1 | - | 3 | 588 | ||
AK296187.1 | - | 4 | 369 | ||
BC013586.1 | - | 3 | 152 | ||
BG323695.1 | - | 565 | |||
BI254784.1 | - | 4 | 550 | ||
J03075.1 | - | 4 | 351 |