Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001007559.2 | NP_001007560.1 | 5 | 560 | |
NM_001308201.1 | NP_001295130.1 | 5 | 535 | ||
NM_005637.3 | NP_005628.2 | 5 | 560 | ||
XM_006722527.2 | XP_006722590.1 | 6 | 586 | ||
XM_011526145.1 | XP_011524447.1 | 5 | 528 | ||
XM_011526147.2 | XP_011524449.1 | 7 | 811 | ||
XM_011526148.2 | XP_011524450.1 | 7 | 798 | ||
XM_011526149.2 | XP_011524451.1 | 7 | 823 | ||
XM_011526150.2 | XP_011524452.1 | 6 | 683 | ||
XM_011526151.2 | XP_011524453.1 | 7 | 812 | ||
XM_011526152.2 | XP_011524454.1 | 6 | 683 | ||
GenBank mRNA | AF244972.1 | - | 5 | 497 | |
AF343880.1 | - | 5 | 494 | ||
AK289510.1 | - | 5 | 538 | ||
AK296949.1 | - | 5 | 518 | ||
AK299082.1 | - | 4 | 428 | ||
AK301504.1 | - | 5 | 565 | ||
AK304305.1 | - | 6 | 676 | ||
AK307923.1 | - | 4 | 451 | ||
AK316302.1 | - | 5 | 510 | ||
AK316338.1 | - | 6 | 629 | ||
BC096221.1 | - | 5 | 497 | 40004908 | |
BC096222.1 | - | 5 | 497 | 40004909 | |
BC096223.1 | - | 5 | 497 | 40004911 | |
BC096224.1 | - | 5 | 497 | 40004912 | |
CR542088.1 | - | 5 | 494 | ||
CR542103.1 | - | 5 | 494 | ||
DC378335.1 | - | 5 | 535 | ||
X79201.1 | - | 5 | 497 |