Gene Transcripts
- Gene Symbol
- MIA
- Entrez Gene ID
- 8190
- Gene Name
- melanoma inhibitory activity
- Gene Aliases
- CD-RAP
- Chromosome Location
- Chr. 19: 40775177 - 40777493 on Build GRCh38
- UniGene ID
- Hs.646364
- Species
- Homo sapiens
- Gene Symbol
- MIA-RAB4B
- Entrez Gene ID
- 100529262
- Gene Name
- MIA-RAB4B readthrough (NMD candidate)
- Gene Aliases
- Chromosome Location
- Chr. 19: 40775537 - 40796944 on Build GRCh38
- UniGene ID
- Hs.631539
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_037775.1 |
|
3 |
299 |
|
GenBank mRNA |
AF087861.1 |
- |
2 |
79 |
|
AK302147.1 |
- |
4 |
715 |
|
View Full Product Details