Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001317800.1 | NP_001304729.1 | 3 | 722 | |
NM_003974.3 | NP_003965.2 | 5 | 1092 | ||
NM_201349.2 | NP_958728.1 | 4 | 1004 | ||
XM_005273680.4 | XP_005273737.1 | 4 | 819 | ||
GenBank mRNA | AF034970.1 | - | 5 | 985 | |
AK226110.1 | - | 3 | 692 | ||
AK292126.1 | - | 5 | 1048 | ||
AK296671.1 | - | 4 | 986 | ||
AL545478.3 | - | 1 | 680 | ||
BC032623.1 | - | 5 | 1060 | 5242017 |