Gene Transcripts
- Gene Symbol
- DHRS2
- Entrez Gene ID
- 10202
- Gene Name
- dehydrogenase/reductase 2
- Gene Aliases
- HEP27, SDR25C1
- Chromosome Location
- Chr. 14: 23630115 - 23645639 on Build GRCh38
- UniGene ID
- Hs.272499
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_005794.3 |
NP_005785.1
|
8, 9 |
1168 |
|
NM_182908.4 |
NP_878912.1
|
8, 9 |
1179 |
|
XM_005267249.1 |
XP_005267306.1
|
8, 9 |
880 |
|
XM_006720001.3 |
XP_006720064.1
|
8, 9 |
843 |
|
XM_011536338.1 |
XP_011534640.1
|
7, 8 |
809 |
|
XM_011536339.2 |
XP_011534641.1
|
7, 8 |
659 |
|
GenBank mRNA |
AB209653.1 |
- |
6, 7 |
4810 |
|
AK092834.1 |
- |
4, 5 |
2579 |
|
AK222857.1 |
- |
8, 9 |
1168 |
|
AK309945.1 |
- |
8, 9 |
2655 |
|
BC002786.2 |
- |
8, 9 |
1170 |
3614599 |
BC007339.2 |
- |
7, 8 |
694 |
|
U31875.1 |
- |
8, 9 |
1154 |
|
View Full Product Details