Gene Transcripts

Gene Symbol
USP24
Entrez Gene ID
23358
Gene Name
ubiquitin specific peptidase 24
Gene Aliases
Chromosome Location
Chr. 1: 55066359 - 55215374 on Build GRCh38
UniGene ID
Hs.477009
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015306.2 NP_056121.2 5 1001
XM_017000831.1 XP_016856320.1 5 1006
XM_017000832.1 XP_016856321.1 5 1003
XM_017000833.1 XP_016856322.1 5 498
XM_017000834.1 XP_016856323.1 5 498
XM_017000835.1 XP_016856324.1 5 1009
XM_017000836.1 XP_016856325.1 5 1009
XM_017000837.1 XP_016856326.1 5 1009

View Full Product Details