Gene Transcripts
- Gene Symbol
- IKZF3
- Entrez Gene ID
- 22806
- Gene Name
- IKAROS family zinc finger 3
- Gene Aliases
- AIO, AIOLOS, ZNFN1A3
- Chromosome Location
- Chr. 17: 39757715 - 39864188 on Build GRCh38
- UniGene ID
- Hs.371680
- Species
- Homo sapiens
- Gene Symbol
- ANAPC16
- Entrez Gene ID
- 119504
- Gene Name
- anaphase promoting complex subunit 16
- Gene Aliases
- APC16, C10orf104, CENP-27, MSAG, bA570G20.3
- Chromosome Location
- Chr. 10: 72216000 - 72235860 on Build GRCh38
- UniGene ID
- Hs.426296
- Species
- Homo sapiens
- Gene Symbol
- ANKRD40
- Entrez Gene ID
- 91369
- Gene Name
- ankyrin repeat domain 40
- Gene Aliases
- Chromosome Location
- Chr. 17: 50693190 - 50707909 on Build GRCh38
- UniGene ID
- Hs.463426
- Species
- Homo sapiens
- Gene Symbol
- CHORDC1
- Entrez Gene ID
- 26973
- Gene Name
- cysteine and histidine rich domain containing 1
- Gene Aliases
- CHP1
- Chromosome Location
- Chr. 11: 90200429 - 90223364 on Build GRCh38
- UniGene ID
- Hs.112482
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK094608.1 |
- |
1 |
640 |
|
- Gene Symbol
- CPT2
- Entrez Gene ID
- 1376
- Gene Name
- carnitine palmitoyltransferase 2
- Gene Aliases
- CPT1, CPTASE, IIAE4
- Chromosome Location
- Chr. 1: 53196429 - 53214197 on Build GRCh38
- UniGene ID
- Hs.713535
- Species
- Homo sapiens
- Gene Symbol
- DKFZP434L187
- Entrez Gene ID
- 26082
- Gene Name
- uncharacterized LOC26082
- Gene Aliases
- Chromosome Location
- Chr. 15: 30196036 - 30214540 on Build GRCh38
- UniGene ID
- Hs.652128
- Species
- Homo sapiens
- Gene Symbol
- DNAPTP3
- Entrez Gene ID
- 100507488
- Gene Name
- histone demethylase UTY-like
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Hs.203717
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AY450391.1 |
- |
1 |
222 |
|
- Gene Symbol
- FARP1
- Entrez Gene ID
- 10160
- Gene Name
- FERM, ARH/RhoGEF and pleckstrin domain protein 1
- Gene Aliases
- CDEP, FARP1-IT1, PLEKHC2, PPP1R75
- Chromosome Location
- Chr. 13: 98142562 - 98449773 on Build GRCh38
- UniGene ID
- Hs.403917
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK123565.1 |
- |
1 |
3690 |
|
- Gene Symbol
- FKBP14
- Entrez Gene ID
- 55033
- Gene Name
- FK506 binding protein 14
- Gene Aliases
- EDSKMH, FKBP22, IPBP12
- Chromosome Location
- Chr. 7: 30010583 - 30026801 on Build GRCh38
- UniGene ID
- Hs.390838
- Species
- Homo sapiens
- Gene Symbol
- GLUL
- Entrez Gene ID
- 2752
- Gene Name
- glutamate-ammonia ligase
- Gene Aliases
- GLNS, GS, PIG43, PIG59
- Chromosome Location
- Chr. 1: 182378093 - 182392206 on Build GRCh38
- UniGene ID
- Hs.132016
- Species
- Homo sapiens
- Gene Symbol
- LINC00094
- Entrez Gene ID
- 266655
- Gene Name
- long intergenic non-protein coding RNA 94
- Gene Aliases
- LP2477, NCRNA00094, bA374P20.3
- Chromosome Location
- Chr. 9: 134025439 - 134031597 on Build GRCh38
- UniGene ID
- Hs.654869
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_015427.2 |
|
3 |
2718 |
|
- Gene Symbol
- LOC105371451
- Entrez Gene ID
- 105371451
- Gene Name
- uncharacterized LOC105371451
- Gene Aliases
- Chromosome Location
- Chr. 1: 155313708 - 155317353 on Build GRCh38
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK125505.1 |
- |
1 |
764 |
|
- Gene Symbol
- LRCH3
- Entrez Gene ID
- 84859
- Gene Name
- leucine rich repeats and calponin homology domain containing 3
- Gene Aliases
- Chromosome Location
- Chr. 3: 197790855 - 197889346 on Build GRCh38
- UniGene ID
- Hs.518414
- Species
- Homo sapiens
- Gene Symbol
- MIPOL1
- Entrez Gene ID
- 145282
- Gene Name
- mirror-image polydactyly 1
- Gene Aliases
- CCDC193
- Chromosome Location
- Chr. 14: 37197888 - 37579207 on Build GRCh38
- UniGene ID
- Hs.660396
- Species
- Homo sapiens
- Gene Symbol
- PKD1L2
- Entrez Gene ID
- 114780
- Gene Name
- polycystin 1 like 2 (gene/pseudogene)
- Gene Aliases
- PC1L2
- Chromosome Location
- Chr. 16: 81100879 - 81220370 on Build GRCh38
- UniGene ID
- Hs.413525
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AL832135.1 |
- |
1 |
2576 |
|
- Gene Symbol
- PLEKHA5
- Entrez Gene ID
- 54477
- Gene Name
- pleckstrin homology domain containing A5
- Gene Aliases
- PEPP-2, PEPP2
- Chromosome Location
- Chr. 12: 19129680 - 19376400 on Build GRCh38
- UniGene ID
- Hs.188614
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC000969.2 |
- |
8 |
2268 |
|
- Gene Symbol
- POLR2F
- Entrez Gene ID
- 5435
- Gene Name
- RNA polymerase II subunit F
- Gene Aliases
- HRBP14.4, POLRF, RPABC14.4, RPABC2, RPB14.4, RPB6, RPC15
- Chromosome Location
- Chr. 22: 37953663 - 38041915 on Build GRCh38
- UniGene ID
- Hs.436578
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK098727.1 |
- |
1 |
1975 |
|
AL832562.1 |
- |
1 |
2210 |
|
- Gene Symbol
- SLC8B1
- Entrez Gene ID
- 80024
- Gene Name
- solute carrier family 8 member B1
- Gene Aliases
- NCKX6, NCLX, SLC24A6
- Chromosome Location
- Chr. 12: 113298766 - 113335142 on Build GRCh38
- UniGene ID
- Hs.286194
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK095092.1 |
- |
1 |
671 |
|
- Gene Symbol
- SNHG20
- Entrez Gene ID
- 654434
- Gene Name
- small nucleolar RNA host gene 20
- Gene Aliases
- C17orf86, LINC00338, NCRNA00338, SCARNA16HG
- Chromosome Location
- Chr. 17: 77088643 - 77094986 on Build GRCh38
- UniGene ID
- Hs.720923
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK098448.1 |
- |
1 |
140 |
|
- Gene Symbol
- SPDYE1
- Entrez Gene ID
- 285955
- Gene Name
- speedy/RINGO cell cycle regulator family member E1
- Gene Aliases
- Ringo1, SPDYE, WBSCR19
- Chromosome Location
- Chr. 7: 43997855 - 44011040 on Build GRCh38
- UniGene ID
- Hs.645483
- Species
- Homo sapiens
- Gene Symbol
- SPDYE2
- Entrez Gene ID
- 441273
- Gene Name
- speedy/RINGO cell cycle regulator family member E2
- Gene Aliases
- SPDYB2-L1
- Chromosome Location
- Chr. 7: 102551232 - 102565227 on Build GRCh38
- UniGene ID
- Hs.656428
- Species
- Homo sapiens
- Gene Symbol
- SPDYE2B
- Entrez Gene ID
- 100310812
- Gene Name
- speedy/RINGO cell cycle regulator family member E2B
- Gene Aliases
- SPDYE2L
- Chromosome Location
- Chr. 7: 102650325 - 102663525 on Build GRCh38
- UniGene ID
- Hs.433879
- Species
- Homo sapiens
- Gene Symbol
- SRSF11P1
- Entrez Gene ID
- 100421296
- Gene Name
- serine and arginine rich splicing factor 11 pseudogene 1
- Gene Aliases
- Chromosome Location
- Chr. 12: 19306671 - 19308236 on Build GRCh38
- UniGene ID
- Hs.655546
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK021937.1 |
- |
1 |
517 |
|
- Gene Symbol
- STT3B
- Entrez Gene ID
- 201595
- Gene Name
- STT3B, catalytic subunit of the oligosaccharyltransferase complex
- Gene Aliases
- CDG1X, SIMP, STT3-B
- Chromosome Location
- Chr. 3: 31532501 - 31637622 on Build GRCh38
- UniGene ID
- Hs.475812
- Species
- Homo sapiens
- Gene Symbol
- TCOF1
- Entrez Gene ID
- 6949
- Gene Name
- treacle ribosome biogenesis factor 1
- Gene Aliases
- MFD1, TCS, TCS1, treacle
- Chromosome Location
- Chr. 5: 150357639 - 150400308 on Build GRCh38
- UniGene ID
- Hs.519672
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AB209317.1 |
- |
13 |
5017 |
|
- Gene Symbol
- TIE1
- Entrez Gene ID
- 7075
- Gene Name
- tyrosine kinase with immunoglobulin like and EGF like domains 1
- Gene Aliases
- JTK14, TIE
- Chromosome Location
- Chr. 1: 43300895 - 43323110 on Build GRCh38
- UniGene ID
- Hs.78824
- Species
- Homo sapiens
- Gene Symbol
- VAPA
- Entrez Gene ID
- 9218
- Gene Name
- VAMP associated protein A
- Gene Aliases
- VAP-33, VAP-A, VAP33, hVAP-33
- Chromosome Location
- Chr. 18: 9913958 - 9960021 on Build GRCh38
- UniGene ID
- Hs.165195
- Species
- Homo sapiens
- Gene Symbol
- ZKSCAN1
- Entrez Gene ID
- 7586
- Gene Name
- zinc finger with KRAB and SCAN domains 1
- Gene Aliases
- 9130423L19Rik, KOX18, PHZ-37, ZNF139, ZNF36, ZSCAN33
- Chromosome Location
- Chr. 7: 100015572 - 100045523 on Build GRCh38
- UniGene ID
- Hs.615360
- Species
- Homo sapiens
- Gene Symbol
- ZNF346-IT1
- Entrez Gene ID
- 106480741
- Gene Name
- ZNF346 intronic transcript 1
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
CV815148.1 |
- |
1 |
19 |
|
- Gene Symbol
- ZNF426
- Entrez Gene ID
- 79088
- Gene Name
- zinc finger protein 426
- Gene Aliases
- K-RBP
- Chromosome Location
- Chr. 19: 9523224 - 9538673 on Build GRCh38
- UniGene ID
- Hs.386476
- Species
- Homo sapiens
- Gene Symbol
- ZNF687
- Entrez Gene ID
- 57592
- Gene Name
- zinc finger protein 687
- Gene Aliases
- PDB6
- Chromosome Location
- Chr. 1: 151281555 - 151291905 on Build GRCh38
- UniGene ID
- Hs.186756
- Species
- Homo sapiens
- Gene Symbol
- ZNHIT3
- Entrez Gene ID
- 9326
- Gene Name
- zinc finger HIT-type containing 3
- Gene Aliases
- TRIP3
- Chromosome Location
- Chr. 17: 36486627 - 36499310 on Build GRCh38
- UniGene ID
- Hs.2210
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AB209193.1 |
- |
1 |
655 |
|
View Full Product Details