Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_018655.2 | NP_061125.1 | 1 | 52 | |
GenBank mRNA | AB451457.1 | - | 1 | 30 | |
AF268478.1 | - | 1 | 52 | ||
BC069140.1 | - | 1 | 52 | 7216999 | |
BC096714.1 | - | 1 | 50 | 40002566 | |
BC098148.1 | - | 1 | 50 | 40002565 | |
BC098246.1 | - | 1 | 50 | 40002556 | |
BC098253.1 | - | 1 | 50 | 40002559 |