Gene Transcripts

Gene Symbol
OSTC
Entrez Gene ID
58505
Gene Name
oligosaccharyltransferase complex non-catalytic subunit
Gene Aliases
DC2
Chromosome Location
Chr. 4: 108650585 - 108667822 on Build GRCh38
UniGene ID
Hs.445803
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001267818.1 NP_001254747.1 2, 3 308
NM_021227.3 NP_067050.1 2, 3 308
GenBank mRNA AF068297.1 - 2, 3 271
AF161425.1 - 2, 3 292
AF201937.1 - 2, 3 299
AF343342.1 - 2, 3 237
AK312185.1 - 2, 3 252
BC016321.1 - 2, 3 296 4075646
BC054857.1 - 2, 3 298 4093030
BM471860.1 - 236
BM541430.1 - 2, 3 259
BM546028.1 - 2, 3 298
BQ435510.1 - 308
CN430159.1 - 2, 3 246

View Full Product Details