Gene Transcripts

Gene Symbol
OR7E91P
Entrez Gene ID
79315
Gene Name
olfactory receptor family 7 subfamily E member 91 pseudogene
Gene Aliases
Chromosome Location
Chr. 2: 71024075 - 71029931 on Build GRCh38
UniGene ID
Hs.327033
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_002185.3 3 699
GenBank mRNA BC014374.1 - 3 702
BC030991.1 - 3 738

View Full Product Details