Gene Transcripts

Gene Symbol
C11orf68
Entrez Gene ID
83638
Gene Name
chromosome 11 open reading frame 68
Gene Aliases
BLES03, P5326
Chromosome Location
Chr. 11: 65916810 - 65919060 on Build GRCh38
UniGene ID
Hs.433573
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135635.1 NP_001129107.1 2 387
NM_031450.3 NP_113638.2 2 384
GenBank mRNA AF073483.1 - 2 293
BC004414.1 - 1 211
BC010512.1 - 1 378 4150027
BC013185.1 - 2 384 4346483
BM705907.1 - 1 247
CX866110.1 - 2 363

View Full Product Details