Gene Transcripts
- Gene Symbol
- DYNLRB2
- Entrez Gene ID
- 83657
- Gene Name
- dynein light chain roadblock-type 2
- Gene Aliases
- DNCL2B, DNLC2B, ROBLD2
- Chromosome Location
- Chr. 16: 80540259 - 80550811 on Build GRCh38
- UniGene ID
- Hs.98849
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001305017.1 |
NP_001291946.1
|
2, 3 |
201 |
|
NM_130897.2 |
NP_570967.1
|
2, 3 |
208 |
|
XM_006721295.3 |
XP_006721358.1
|
2, 3 |
405 |
|
XM_006721296.3 |
XP_006721359.1
|
2, 3 |
158 |
|
XM_011523370.2 |
XP_011521672.1
|
2, 3 |
165 |
|
GenBank mRNA |
AF125108.1 |
- |
2, 3 |
203 |
|
CD676360.1 |
- |
2, 3 |
201 |
|
DB458006.1 |
- |
2, 3 |
170 |
|
HY018370.1 |
- |
2, 3 |
209 |
|
HY123054.1 |
- |
2, 3 |
191 |
|
HY201608.1 |
- |
2, 3 |
456 |
|
View Full Product Details