Gene Transcripts
- Gene Symbol
- TRIM52
- Entrez Gene ID
- 84851
- Gene Name
- tripartite motif containing 52
- Gene Aliases
- RNF102
- Chromosome Location
- Chr. 5: 181254077 - 181261160 on Build GRCh38
- UniGene ID
- Hs.458412
- Species
- Homo sapiens
- Gene Symbol
- CTC-338M12.4
- Entrez Gene ID
- 101928649
- Gene Name
- uncharacterized LOC101928649
- Gene Aliases
- Chromosome Location
- Chr. 5: 181246541 - 181257586 on Build GRCh38
- UniGene ID
- Hs.723307
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_109909.1 |
|
3 |
896 |
|
View Full Product Details