Gene Transcripts
- Gene Symbol
- LRR1
- Entrez Gene ID
- 122769
- Gene Name
- leucine rich repeat protein 1
- Gene Aliases
- 4-1BBLRR, LRR-1, PPIL5
- Chromosome Location
- Chr. 14: 49598697 - 49614672 on Build GRCh38
- UniGene ID
- Hs.451090
- Species
- Homo sapiens
- Gene Symbol
- LOC100288910
- Entrez Gene ID
- 100288910
- Gene Name
- uncharacterized LOC100288910
- Gene Aliases
- Chromosome Location
- Chr. 14: 49597660 - 49599158 on Build GRCh38
- UniGene ID
- Hs.156367
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BX161511.1 |
- |
1 |
126 |
|
View Full Product Details