Gene Transcripts
- Gene Symbol
- FAM213B
- Entrez Gene ID
- 127281
- Gene Name
- family with sequence similarity 213 member B
- Gene Aliases
- C1orf93
- Chromosome Location
- Chr. 1: 2586460 - 2591469 on Build GRCh38
- UniGene ID
- Hs.462033
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001195737.1 |
NP_001182666.1
|
3, 4 |
450 |
|
NM_001195738.1 |
NP_001182667.1
|
3, 4 |
450 |
|
NM_001195740.1 |
NP_001182669.1
|
3, 4 |
450 |
|
NM_001195741.1 |
NP_001182670.1
|
3, 4 |
450 |
|
NM_152371.3 |
NP_689584.2
|
3, 4 |
450 |
|
NR_036637.1 |
|
3, 4 |
383 |
|
NR_036638.1 |
|
3, 4 |
383 |
|
XM_006710354.3 |
XP_006710417.1
|
3, 4 |
418 |
|
GenBank mRNA |
AF425266.1 |
- |
3, 4 |
394 |
|
AK075273.1 |
- |
3, 4 |
402 |
|
AK094901.1 |
- |
2, 3 |
264 |
|
AK291908.1 |
- |
3, 4 |
408 |
|
AK298926.1 |
- |
3, 4 |
404 |
|
AK303504.1 |
- |
3, 4 |
422 |
|
AK308034.1 |
- |
3, 4 |
360 |
|
AK316243.1 |
- |
3, 4 |
360 |
|
BC022547.1 |
- |
3, 4 |
390 |
4812314 |
BI908668.1 |
- |
3, 4 |
302 |
|
BQ709693.1 |
- |
|
286 |
|
DA292832.1 |
- |
3, 4 |
382 |
|
DA495138.1 |
- |
3, 4 |
383 |
|
DC351872.1 |
- |
3, 4 |
425 |
|
DC407959.1 |
- |
3, 4 |
450 |
|
View Full Product Details