Gene Transcripts

Gene Symbol
NEU4
Entrez Gene ID
129807
Gene Name
neuraminidase 4
Gene Aliases
Chromosome Location
Chr. 2: 241809065 - 241817413 on Build GRCh38
UniGene ID
Hs.551747
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001167599.2 NP_001161071.1 4 1408
NM_001167600.2 NP_001161072.1 4 1085
NM_001167601.2 NP_001161073.1 4 1088
NM_001167602.2 NP_001161074.1 5 1586
NM_080741.3 NP_542779.2 4 1405
GenBank mRNA AJ277883.1 - 3 872
AK025617.1 - 5 1338
AK091038.1 - 5 1583
AK096992.1 - 2 2198
AK289421.1 - 4 962
AK290076.1 - 4 945
AK298403.1 - 5 917
AK300969.1 - 4 958
BC012899.1 - 4 1359 4156395
BC105974.1 - 4 921 4944243
BC117214.1 - 4 1049 40125765

View Full Product Details