Gene Transcripts
- Gene Symbol
- MSL3P1
- Entrez Gene ID
- 151507
- Gene Name
- male-specific lethal 3 homolog (Drosophila) pseudogene 1
- Gene Aliases
- MSL3L2
- Chromosome Location
- Chr. 2: 233865437 - 233868409 on Build GRCh38
- UniGene ID
- Hs.355809
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_024322.1 |
|
2 |
1036 |
|
GenBank mRNA |
AK301707.1 |
- |
2 |
1020 |
|
AK316372.1 |
- |
2 |
987 |
|
View Full Product Details