Gene Transcripts

Gene Symbol
FAM65C
Entrez Gene ID
140876
Gene Name
family with sequence similarity 65 member C
Gene Aliases
C20orf175, C20orf176
Chromosome Location
Chr. 20: 50586108 - 50692004 on Build GRCh38
UniGene ID
Hs.372578
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001290268.1 NP_001277197.1 22 3238
NM_080829.3 NP_543019.2 22 3171
NR_110890.1 22 3397
XM_005260294.3 XP_005260351.1 23 3235
XM_006723713.3 XP_006723776.1 22 3283
XM_011528578.2 XP_011526880.1 23 3263
XM_011528579.2 XP_011526881.1 23 3401
XM_011528580.2 XP_011526882.1 23 3455
XM_011528581.2 XP_011526883.1 23 3439
XM_011528585.2 XP_011526887.1 12 2586
XM_017027681.1 XP_016883170.1 14 2231
GenBank mRNA AK299337.1 - 22 3087
CA389799.1 - 3 223
CD558804.1 - 3 265

View Full Product Details