Gene Transcripts

Gene Symbol
NUDT14
Entrez Gene ID
256281
Gene Name
nudix hydrolase 14
Gene Aliases
UGPP, UGPPase
Chromosome Location
Chr. 14: 105172937 - 105181323 on Build GRCh38
UniGene ID
Hs.526432
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177533.4 NP_803877.2 5 567
GenBank mRNA AB087802.1 - 5 453
AK128196.1 - 7 3032
BC041584.1 - 5 529 4810207
BE671270.1 - 1 301

View Full Product Details