Gene Transcripts

Gene Symbol
CRACR2B
Entrez Gene ID
283229
Gene Name
calcium release activated channel regulator 2B
Gene Aliases
EFCAB4A
Chromosome Location
Chr. 11: 825558 - 831991 on Build GRCh38
UniGene ID
Hs.660936
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001286606.1 NP_001273535.1 4 874
NM_173584.4 NP_775855.3 4 874
XM_011520011.1 XP_011518313.1 4 1456
XM_011520019.1 XP_011518321.1 4 1456
XM_017017581.1 XP_016873070.1 4 1456
XM_017017582.1 XP_016873071.1 5 1263
XM_017017583.1 XP_016873072.1 5 1235
XM_017017584.1 XP_016873073.1 5 1275
XM_017017585.1 XP_016873074.1 5 1814
XM_017017586.1 XP_016873075.1 5 1135
XM_017017587.1 XP_016873076.1 5 1506
XM_017017588.1 XP_016873077.1 5 930
XM_017017589.1 XP_016873078.1 4 1438
XM_017017590.1 XP_016873079.1 5 1045
XM_017017591.1 XP_016873080.1 5 1015
XM_017017592.1 XP_016873081.1 5 1594
XM_017017593.1 XP_016873082.1 5 915
XM_017017594.1 XP_016873083.1 5 1286
XM_017017595.1 XP_016873084.1 5 710
XM_017017596.1 XP_016873085.1 4 1218
XM_017017597.1 XP_016873086.1 4 1456
XM_017017598.1 XP_016873087.1 4 1456
XM_017017599.1 XP_016873088.1 5 1113
GenBank mRNA AK075180.1 - 2 330
AK097334.1 - 1 747
AK127926.1 - 2 304
BC033196.1 - 4 655 4849453
BC073821.1 - 5 968 5952348
GU952799.1 - 4 526

View Full Product Details