Gene Transcripts

Gene Symbol
SSC5D
Entrez Gene ID
284297
Gene Name
scavenger receptor cysteine rich family member with 5 domains
Gene Aliases
S5D-SRCRB
Chromosome Location
Chr. 19: 55488503 - 55519099 on Build GRCh38
UniGene ID
Hs.554182
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001144950.1 NP_001138422.1 14 4614
GenBank mRNA AF086519.1 - 1 138
BC130538.1 - 1 106
BC130540.1 - 1 106
EU699476.1 - 14 4550

View Full Product Details