Gene Transcripts

Gene Symbol
KRTAP20-1
Entrez Gene ID
337975
Gene Name
keratin associated protein 20-1
Gene Aliases
KAP20.1
Chromosome Location
Chr. 21: 30616455 - 30616625 on Build GRCh38
UniGene ID
Hs.553697
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181615.1 NP_853646.1 1 87
GenBank mRNA AB096955.1 - 1 117
BC104846.1 - 1 111 8143849
BC104872.1 - 1 111 8143875

View Full Product Details