Gene Transcripts

Gene Symbol
ANO9
Entrez Gene ID
338440
Gene Name
anoctamin 9
Gene Aliases
PIG5, TMEM16J, TP53I5
Chromosome Location
Chr. 11: 417930 - 442011 on Build GRCh38
UniGene ID
Hs.501622
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012302.2 NP_001012302.2 20 1998
XM_011520053.2 XP_011518355.1 20 2094
GenBank mRNA AK026923.1 - 20 1945
AK074088.1 - 13 4615
AK096874.1 - 11 1286
AL832289.1 - 9 1920
BC047946.1 - 1 599
BC128557.1 - 20 1966 40122929
BC144415.1 - 19 2024

View Full Product Details