Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001012241.1 | NP_001012241.1 | 3 | 405 | |
XM_005257298.4 | XP_005257355.1 | 2 | 2057 | ||
XM_005257299.4 | XP_005257356.3 | 2 | 2057 | ||
GenBank mRNA | BC118997.1 | - | 3 | 328 | |
BC122543.1 | - | 3 | 328 | 40084757 | |
CR749360.1 | - | 3 | 356 |