Gene Transcripts

Gene Symbol
NBPF7
Entrez Gene ID
343505
Gene Name
neuroblastoma breakpoint family member 7
Gene Aliases
Chromosome Location
Chr. 1: 119833730 - 119844880 on Build GRCh38
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001047980.2 NP_001041445.1 7 1312
XM_017001186.1 XP_016856675.1 6 912

View Full Product Details