Gene Transcripts
- Gene Symbol
- DPY19L2P2
- Entrez Gene ID
- 349152
- Gene Name
- DPY19L2 pseudogene 2
- Gene Aliases
- Chromosome Location
- Chr. 7: 103175013 - 103280466 on Build GRCh38
- UniGene ID
- Hs.732579
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_003561.2 |
|
7, 8 |
2203 |
|
GenBank mRNA |
AK131353.1 |
- |
7, 8 |
2196 |
|
View Full Product Details