Gene Transcripts

Gene Symbol
NEO1
Entrez Gene ID
4756
Gene Name
neogenin 1
Gene Aliases
IGDCC2, NGN, NTN1R2
Chromosome Location
Chr. 15: 73051715 - 73305206 on Build GRCh38
UniGene ID
Hs.388613
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001172623.1 NP_001166094.1 28 6750
NM_001172624.1 NP_001166095.1 28 6876
NM_002499.3 NP_002490.2 29 6909
XM_005254408.1 XP_005254465.1 30 7219
XM_011521628.2 XP_011519930.1 29 6974
XM_011521629.2 XP_011519931.1 28 6935
XM_011521630.2 XP_011519932.1 29 6920
XM_011521632.2 XP_011519934.1 27 6866
XM_011521633.2 XP_011519935.1 28 6809
XM_017022231.1 XP_016877720.1 28 6827
XM_017022232.1 XP_016877721.1 29 7138
XM_017022233.1 XP_016877722.1 27 6776
XM_017022234.1 XP_016877723.1 29 6804
XM_017022235.1 XP_016877724.1 27 6728
XM_017022236.1 XP_016877725.1 27 6716
XM_017022237.1 XP_016877726.1 26 6707
XM_017022238.1 XP_016877727.1 27 6668
GenBank mRNA AB209412.1 - 24 5724
AL355708.1 - 1 1517
AL359597.1 - 1 1079
AL390182.1 - 1 1517

View Full Product Details