Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_021965.3 | NP_068800.2 | 4 | 853 | |
XM_011518782.2 | XP_011517084.1 | 4 | 1143 | ||
XM_011518783.2 | XP_011517085.1 | 4 | 1143 | ||
XM_017014808.1 | XP_016870297.1 | 4 | 745 | ||
GenBank mRNA | AK297108.1 | - | 4 | 675 | |
BC033073.1 | - | 4 | 853 | 4123706 |