Gene Transcripts
- Gene Symbol
- KLF17
- Entrez Gene ID
- 128209
- Gene Name
- Kruppel like factor 17
- Gene Aliases
- ZNF393, Zfp393
- Chromosome Location
- Chr. 1: 44043927 - 44135140 on Build GRCh38
- UniGene ID
- Hs.567674
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC031250.1 |
- |
7 |
1502 |
|
View Full Product Details