Gene Transcripts

Gene Symbol
WDR27
Entrez Gene ID
253769
Gene Name
WD repeat domain 27
Gene Aliases
Chromosome Location
Chr. 6: 169426420 - 169702071 on Build GRCh38
UniGene ID
Hs.131903
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_011535687.2 XP_011533989.1 27 5619
XM_011535688.2 XP_011533990.1 27 5615
XM_017010661.1 XP_016866150.1 27 5619

View Full Product Details