Gene Transcripts
- Gene Symbol
- KPNA7
- Entrez Gene ID
- 402569
- Gene Name
- karyopherin subunit alpha 7
- Gene Aliases
- IPOA8
- Chromosome Location
- Chr. 7: 99145467 - 99219417 on Build GRCh38
- UniGene ID
- Hs.253050
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
XM_011516215.2 |
XP_011514517.1
|
6, 7 |
995 |
|
XM_017012208.1 |
XP_016867697.1
|
6, 7 |
995 |
|
XM_017012209.1 |
XP_016867698.1
|
6, 7 |
983 |
|
XM_017012210.1 |
XP_016867699.1
|
6, 7 |
1071 |
|
XM_017012211.1 |
XP_016867700.1
|
6, 7 |
995 |
|
XM_017012212.1 |
XP_016867701.1
|
6, 7 |
785 |
|
XM_017012213.1 |
XP_016867702.1
|
6, 7 |
1123 |
|
XM_017012214.1 |
XP_016867703.1
|
6, 7 |
699 |
|
XM_017012215.1 |
XP_016867704.1
|
6, 7 |
995 |
|
XM_017012216.1 |
XP_016867705.1
|
3, 4 |
443 |
|
GenBank mRNA |
EU126604.1 |
- |
5, 6 |
639 |
|
View Full Product Details