Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_017741.3 | NP_060211.3 | 3 | 458 | |
XM_005248169.3 | XP_005248226.1 | 3 | 446 | ||
XM_005248171.3 | XP_005248228.1 | 2 | 342 | ||
XM_006713966.3 | XP_006714029.1 | 4 | 617 | ||
XM_017008327.1 | XP_016863816.1 | 2 | 1911 | ||
XM_017008328.1 | XP_016863817.1 | 2 | 1896 | ||
GenBank mRNA | AK056116.1 | - | 3 | 458 | |
BC050697.1 | - | 2 | 119 | 6044417 |