Gene Transcripts

Gene Symbol
SLC37A3
Entrez Gene ID
84255
Gene Name
solute carrier family 37 member 3
Gene Aliases
Chromosome Location
Chr. 7: 140333752 - 140404352 on Build GRCh38
UniGene ID
Hs.446021
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_011516626.2 XP_011514928.1 2 345
XM_011516627.2 XP_011514929.1 2 344
XM_017012712.1 XP_016868201.1 2 321
XM_017012713.1 XP_016868202.1 2 340
XM_017012714.1 XP_016868203.1 2 361
XM_017012715.1 XP_016868204.1 2 364
XM_017012716.1 XP_016868205.1 2 368
XM_017012717.1 XP_016868206.1 2 340
XM_017012718.1 XP_016868207.1 2 339
GenBank mRNA AK131347.1 - 2 449

View Full Product Details