Gene Transcripts
- Gene Symbol
- FAM86C2P
- Entrez Gene ID
- 645332
- Gene Name
- family with sequence similarity 86, member A pseudogene
- Gene Aliases
- Chromosome Location
- Chr. 11: 67791767 - 67805336 on Build GRCh38
- UniGene ID
- Hs.535094
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK055771.1 |
- |
1 |
309 |
|
- Gene Symbol
- FAM86EP
- Entrez Gene ID
- 348926
- Gene Name
- family with sequence similarity 86, member A pseudogene
- Gene Aliases
- Chromosome Location
- Chr. 4: 3941760 - 3955428 on Build GRCh38
- UniGene ID
- Hs.591720
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_130740.1 |
|
3 |
948 |
|
View Full Product Details