Gene Transcripts

Gene Symbol
KPRP
Entrez Gene ID
448834
Gene Name
keratinocyte proline rich protein
Gene Aliases
C1orf45
Chromosome Location
Chr. 1: 152758030 - 152762053 on Build GRCh38
UniGene ID
Hs.149386
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025231.1 NP_001020402.1 2 540
XM_011509571.1 XP_011507873.1 2 581
GenBank mRNA AY960854.1 - 1 511

View Full Product Details