Gene Transcripts

Gene Symbol
PRDM11
Entrez Gene ID
56981
Gene Name
PR domain 11
Gene Aliases
PFM8
Chromosome Location
Chr. 11: 45094013 - 45235124 on Build GRCh38
UniGene ID
Hs.178715
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_011520230.2 XP_011518532.1 7 2090
XM_017018024.1 XP_016873513.1 5 1393
GenBank mRNA AK097878.1 - 2 334
AY726563.1 - 2 1041

Gene Symbol
LOC101060179
Entrez Gene ID
101060179
Gene Name
uncharacterized LOC101060179
Gene Aliases
Chromosome Location
Chr. 11: 45213067 - 45214744 on Build GRCh38
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003959932.4 XP_003959981.1 1 138

View Full Product Details