Gene Transcripts
- Gene Symbol
- RNASET2
- Entrez Gene ID
- 8635
- Gene Name
- ribonuclease T2
- Gene Aliases
- RNASE6PL, bA514O12.3
- Chromosome Location
- Chr. 6: 166929509 - 166956589 on Build GRCh38
- UniGene ID
- Hs.529989
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK124363.1 |
- |
1 |
225 |
|
View Full Product Details