Gene Transcripts

Gene Symbol
UBE2Q2P2
Entrez Gene ID
100134869
Gene Name
ubiquitin conjugating enzyme E2 Q2 pseudogene 2
Gene Aliases
UBE2Q2P3, UBE2QP2
Chromosome Location
Chr. 15: 82355136 - 82415611 on Build GRCh38
UniGene ID
Hs.735222
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_004847.3 2, 3 285
GenBank mRNA BI916968.1 - 2, 3 304
CN343524.1 - 2, 3 273
DA194105.1 - 2, 3 279
DA527368.1 - 2, 3 303
DA778756.1 - 2, 3 354

View Full Product Details