Gene Transcripts

Gene Symbol
PKN3
Entrez Gene ID
29941
Gene Name
protein kinase N3
Gene Aliases
UTDP4-1
Chromosome Location
Chr. 9: 128702497 - 128720920 on Build GRCh38
UniGene ID
Hs.300485
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_005251946.3 XP_005252003.1 1 556
GenBank mRNA AK097535.1 - 1 543

View Full Product Details